Assay Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 14 - 15 | 1829 | 90 | |
| GenBank mRNA | AK147084.1 | - | 14 - 15 | 1830 | 90 | |
| AK163950.1 | - | 4 - 5 | 517 | 90 | ||
| BC096493.1 | - | 14 - 15 | 1623 | 30742571 | 90 | |
| M25811.1 | - | 14 - 15 | 1863 | 90 | ||
| X52684.1 | - | 15 - 16 | 1770 | 90 | ||
| X52685.1 | - | 15 - 16 | 1770 | 90 | ||
More Information
Set Membership: |
|

Prkca
Genomic Map