Assay Details
| Gene Symbol: Prkca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011101.3 | NP_035231.2 | 3 - 4 | 510 | 89 | |
| GenBank mRNA | AK147084.1 | - | 3 - 4 | 511 | 89 | |
| BC096493.1 | - | 3 - 4 | 304 | 30742571 | 89 | |
| M25811.1 | - | 3 - 4 | 544 | 89 | ||
| X52685.1 | - | 4 - 5 | 451 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkca
Genomic Map