Assay Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112705.2 | NP_001106176.1 | 12 - 13 | 1647 | 125 | |
| NM_001294329.1 | NP_001281258.1 | 10 - 11 | 1507 | 125 | ||
| NM_001294330.1 | NP_001281259.1 | 10 - 11 | 1455 | 125 | ||
| NM_001294331.1 | NP_001281260.1 | 12 - 13 | 1660 | 125 | ||
| NM_001294334.1 | NP_001281263.1 | 12 - 13 | 1647 | 125 | ||
| GenBank mRNA | AF045252.1 | - | 12 - 13 | 1666 | 125 | |
| AF045253.1 | - | 11 - 12 | 1094 | 125 | ||
| AF045254.1 | - | 7 - 8 | 808 | 125 | ||
| AK014829.1 | - | 12 - 13 | 1285 | 125 | ||
| AK089925.1 | - | 10 - 11 | 1035 | 125 | ||
| AK165667.1 | - | 12 - 13 | 1650 | 125 | ||
| AX775743.1 | - | 6 - 7 | 672 | 125 | ||
| BC066198.1 | - | 10 - 11 | 1000 | 125 | ||
| U56909.1 | - | 3 - 4 | 345 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlk2
Genomic Map