Assay Details
| Gene Symbol: Tlk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112705.2 | NP_001106176.1 | 19 - 20 | 2388 | 65 | |
| NM_001294329.1 | NP_001281258.1 | 17 - 18 | 2248 | 65 | ||
| NM_001294330.1 | NP_001281259.1 | 17 - 18 | 2196 | 65 | ||
| NM_001294331.1 | NP_001281260.1 | 19 - 20 | 2401 | 65 | ||
| NM_001294334.1 | NP_001281263.1 | 19 - 20 | 2388 | 65 | ||
| GenBank mRNA | AF045252.1 | - | 19 - 20 | 2407 | 65 | |
| AF045253.1 | - | 18 - 19 | 1835 | 65 | ||
| AF045254.1 | - | 14 - 15 | 1549 | 65 | ||
| AK014829.1 | - | 19 - 20 | 2026 | 65 | ||
| AK161383.1 | - | 1 - 2 | 156 | 65 | ||
| AK165667.1 | - | 19 - 20 | 2391 | 65 | ||
| AX775743.1 | - | 13 - 14 | 1413 | 65 | ||
| BC066198.1 | - | 17 - 18 | 1741 | 65 | ||
| U56909.1 | - | 10 - 11 | 1086 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlk2
Genomic Map