Assay Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033327.2 | NP_201584.2 | 5 - 6 | 3959 | 60 | |
| GenBank mRNA | AF188609.1 | - | 3 - 4 | 3328 | 60 | |
| AK122365.1 | - | 5 - 6 | 3925 | 60 | ||
| AK133497.1 | - | 3 - 4 | 3387 | 60 | ||
| AY147407.1 | - | 4 - 5 | 3564 | 60 | ||
| AY256893.1 | - | 5 - 6 | 3858 | 60 | ||
| BC059234.1 | - | 3 - 4 | 3770 | 60 | ||
| BC079586.1 | - | 5 - 6 | 3645 | 60 | ||
| BC139028.1 | - | 5 - 6 | 3705 | 8862050 | 60 | |
| BC139030.1 | - | 5 - 6 | 3705 | 8862052 | 60 | |
| BU519630.1 | - | 2 - 3 | 182 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp423
Genomic Map