Assay Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001445.2 | NP_001001445.1 | 13 - 14 | 2356 | 67 | |
| GenBank mRNA | AB180097.1 | - | 12 - 13 | 2104 | 67 | |
| AJ620495.1 | - | 12 - 13 | 2144 | 67 | ||
| AK051315.1 | - | 13 - 14 | 2214 | 67 | ||
| AY445519.1 | - | 12 - 13 | 2104 | 67 | ||
| AY452083.1 | - | 12 - 13 | 2137 | 67 | ||
| AY452084.1 | - | 12 - 13 | 2107 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Trpv1
Genomic Map