Assay Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001445.2 | NP_001001445.1 | 10 - 11 | 1808 | 68 | |
| GenBank mRNA | AB180097.1 | - | 9 - 10 | 1556 | 68 | |
| AJ620495.1 | - | 9 - 10 | 1596 | 68 | ||
| AK051315.1 | - | 10 - 11 | 1666 | 68 | ||
| AY445519.1 | - | 9 - 10 | 1556 | 68 | ||
| AY452083.1 | - | 9 - 10 | 1589 | 68 | ||
| AY452084.1 | - | 9 - 10 | 1559 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpv1
Genomic Map