Assay Details
| Gene Symbol: Grb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008163.3 | NP_032189.1 | 1 - 2 | 202 | 53 | |
| GenBank mRNA | AK148148.1 | - | 1 - 2 | 167 | 53 | |
| AK166142.1 | - | 1 - 2 | 156 | 53 | ||
| AK172002.1 | - | 1 - 2 | 177 | 53 | ||
| AK196759.1 | - | 1 - 2 | 26 | 53 | ||
| BC052377.1 | - | 1 - 2 | 202 | 5696248 | 53 | |
| BC085254.1 | - | 1 - 2 | 38 | 5343274 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148148.1, AK166142.1, BC052377.1, BC085254.1, NM_008163.3 transcripts.
Set Membership: |
|

Grb2
Genomic Map