Assay Details
| Gene Symbol: Rab37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163753.1 | NP_001157225.1 | 2 - 3 | 612 | 70 | |
| GenBank mRNA | AK045995.1 | - | 2 - 3 | 215 | 70 | |
| AK046260.1 | - | 2 - 3 | 613 | 70 | ||
More Information
Set Membership: |
|

Rab37
Genomic Map