Assay Details
| Gene Symbol: Rab37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163753.1 | NP_001157225.1 | 6 - 7 | 843 | 93 | |
| NM_021411.4 | NP_067386.3 | 6 - 7 | 455 | 93 | ||
| GenBank mRNA | AB232633.1 | - | 6 - 7 | 434 | 93 | |
| AF233582.1 | - | 6 - 7 | 434 | 93 | ||
| AK039716.1 | - | 6 - 7 | 449 | 93 | ||
| AK040216.1 | - | 6 - 7 | 455 | 93 | ||
| AK045995.1 | - | 6 - 7 | 446 | 93 | ||
| AK046260.1 | - | 6 - 7 | 844 | 93 | ||
| AK050483.1 | - | 6 - 7 | 449 | 93 | ||
| BC050757.1 | - | 6 - 7 | 455 | 93 | ||
| BC128020.1 | - | 6 - 7 | 453 | 40136290 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab37
Genomic Map