Assay Details
| Gene Symbol: Slc9a3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012030.2 | NP_036160.1 | 2 - 3 | 771 | 91 | |
| GenBank mRNA | AK085210.1 | - | 2 - 3 | 773 | 91 | |
| AK153558.1 | - | 2 - 3 | 771 | 91 | ||
| AK168894.1 | - | 2 - 3 | 772 | 91 | ||
| BC085141.1 | - | 2 - 3 | 756 | 6413140 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a3r1
Genomic Map