Assay Details
| Gene Symbol: Ube2o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173755.3 | NP_776116.2 | 13 - 14 | 2137 | 64 | |
| GenBank mRNA | AK129431.1 | - | 13 - 14 | 2137 | 64 | |
| BC039233.1 | - | 5 - 6 | 603 | 64 | ||
| BC051108.1 | - | 4 - 5 | 482 | 64 | ||
| BC059193.1 | - | 13 - 14 | 1762 | 4160667 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2o
Genomic Map