Assay Details
| Gene Symbol: Ube2o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173755.3 | NP_776116.2 | 1 - 2 | 435 | 84 | |
| GenBank mRNA | AK129431.1 | - | 1 - 2 | 435 | 84 | |
| BC059193.1 | - | 1 - 2 | 60 | 4160667 | 84 | |
| U20780.1 | - | 1 - 2 | 97 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059193.1 transcripts.
Set Membership: |
|

Ube2o
Genomic Map