Assay Details
| Gene Symbol: St6galnac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009180.3 | NP_033206.2 | 2 - 3 | 285 | 60 | |
| GenBank mRNA | AK004613.1 | - | 2 - 3 | 286 | 60 | |
| AK136381.1 | - | 288 | 60 | |||
| BC010208.1 | - | 237 | 60 | |||
| X93999.1 | - | 2 - 3 | 249 | 60 | ||
More Information
Set Membership: |
|

St6galnac2
Genomic Map