Assay Details
| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162872.1 | NP_001156344.1 | 12 - 13 | 1469 | 82 | |
| NM_016857.2 | NP_058553.2 | 14 - 15 | 1601 | 82 | ||
| GenBank mRNA | AF014461.1 | - | 14 - 15 | 1536 | 82 | |
| AK087601.1 | - | 12 - 13 | 1457 | 82 | ||
| AK140450.1 | - | 14 - 15 | 1575 | 82 | ||
| AK167769.1 | - | 12 - 13 | 1452 | 82 | ||
| AK169928.1 | - | 12 - 13 | 1456 | 82 | ||
| AK173094.1 | - | 12 - 13 | 6088 | 82 | ||
| BC028927.1 | - | 12 - 13 | 1445 | 4208803 | 82 | |
| BC053710.1 | - | 12 - 13 | 1456 | 30056570 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc7
Genomic Map