Assay Details
| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162872.1 | NP_001156344.1 | 14 - 15 | 1687 | 70 | |
| NM_016857.2 | NP_058553.2 | 16 - 17 | 1819 | 70 | ||
| GenBank mRNA | AF014461.1 | - | 16 - 17 | 1754 | 70 | |
| AK087601.1 | - | 14 - 15 | 1675 | 70 | ||
| AK140450.1 | - | 16 - 17 | 1793 | 70 | ||
| AK167769.1 | - | 14 - 15 | 1670 | 70 | ||
| AK169928.1 | - | 14 - 15 | 1674 | 70 | ||
| AK173094.1 | - | 14 - 15 | 6306 | 70 | ||
| BC028927.1 | - | 14 - 15 | 1663 | 4208803 | 70 | |
| BC053710.1 | - | 14 - 15 | 1674 | 30056570 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc7
Genomic Map