Assay Details
| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162872.1 | NP_001156344.1 | 1 - 2 | 122 | 64 | |
| NM_016857.2 | NP_058553.2 | 1 - 2 | 122 | 64 | ||
| GenBank mRNA | AK087601.1 | - | 1 - 2 | 110 | 64 | |
| AK140450.1 | - | 1 - 2 | 96 | 64 | ||
| AK162868.1 | - | 1 - 2 | 108 | 64 | ||
| AK167769.1 | - | 1 - 2 | 105 | 64 | ||
| AK173094.1 | - | 1 - 2 | 98 | 64 | ||
| BC053710.1 | - | 1 - 2 | 109 | 30056570 | 64 | |
| BP757578.1 | - | 1 - 2 | 149 | 64 | ||
More Information
Set Membership: |
|

Exoc7
Genomic Map