| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100038902.mm.1 |
NM_001162872.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038902.mm.1 |
NM_001162872.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038902.mm.1 |
NM_016857.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038902.mm.1 |
NM_016857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038902.mm.1 |
TC1100004113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038904.mm.1 |
NM_001162872.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038904.mm.1 |
NM_001162872.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038904.mm.1 |
NM_016857.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038904.mm.1 |
NM_016857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038904.mm.1 |
TC1100004113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038900.mm.1 |
NM_001162872.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038900.mm.1 |
NM_001162872.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038900.mm.1 |
NM_016857.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038900.mm.1 |
NM_016857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038900.mm.1 |
TC1100004113.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100038903.mm.1 |
NM_001162872.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038903.mm.1 |
NM_001162872.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038903.mm.1 |
NM_016857.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038903.mm.1 |
NM_016857.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100038903.mm.1 |
TC1100004113.mm.1 |
MTA TC |