Assay Details
| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162872.1 | NP_001156344.1 | 9 - 10 | 1274 | 88 | |
| NM_016857.2 | NP_058553.2 | 10 - 11 | 1367 | 88 | ||
| GenBank mRNA | AK087601.1 | - | 9 - 10 | 1262 | 88 | |
| AK140450.1 | - | 10 - 11 | 1341 | 88 | ||
| AK162868.1 | - | 9 - 10 | 1687 | 88 | ||
| AK167769.1 | - | 9 - 10 | 1257 | 88 | ||
| AK169928.1 | - | 9 - 10 | 1261 | 88 | ||
| AK173094.1 | - | 8 - 9 | 5854 | 88 | ||
| BC028927.1 | - | 9 - 10 | 1250 | 4208803 | 88 | |
| BC053710.1 | - | 9 - 10 | 1261 | 30056570 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exoc7
Genomic Map