Assay Details
| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011150.2 | NP_035280.1 | 2 - 3 | 216 | 61 | |
| GenBank mRNA | AK079915.1 | - | 2 - 3 | 245 | 61 | |
| AK137836.1 | - | 2 - 3 | 193 | 61 | ||
| AK150021.1 | - | 2 - 3 | 245 | 61 | ||
| AK152182.1 | - | 2 - 3 | 158 | 61 | ||
| AK153313.1 | - | 2 - 3 | 232 | 61 | ||
| AK153396.1 | - | 2 - 3 | 234 | 61 | ||
| AK155031.1 | - | 2 - 3 | 158 | 61 | ||
| BC090658.1 | - | 2 - 3 | 212 | 30666981 | 61 | |
| X67809.1 | - | 2 - 3 | 216 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lgals3bp
Genomic Map