Assay Details
| Gene Symbol: Cant1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267592.1 | NP_001254521.1 | 4 - 5 | 1040 | 115 | |
| GenBank mRNA | AK006565.1 | - | 4 - 5 | 891 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cant1
Genomic Map