Assay Details
| Gene Symbol: Syngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009304.2 | NP_033330.1 | 3 - 4 | 512 | 84 | |
| GenBank mRNA | AF151985.1 | - | 3 - 4 | 509 | 84 | |
| AF364049.1 | - | 3 - 4 | 492 | 84 | ||
| AJ002307.1 | - | 3 - 4 | 478 | 84 | ||
| AK150234.1 | - | 3 - 4 | 512 | 84 | ||
| AK170681.1 | - | 3 - 4 | 512 | 84 | ||
| BC004829.1 | - | 3 - 4 | 505 | 2646657 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28167341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28167340 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106235 | AK157343.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106235 | AK170681.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106235 | AK150234.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106235 | AF151985.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2106235 | NM_009304.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 572156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628050 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125505 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106235 | AK157343.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106235 | AK170681.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106235 | AK150234.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106235 | AF151985.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2106235 | NM_009304.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167345 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167333 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167319 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167340 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167330 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167338 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167346 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167337 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167315 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167336 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167324 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167322 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167334 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167343 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167331 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167349 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167348 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28167335 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125505 | - | - |
| Whole Mouse Genome G4122A | A_51_P105986 | AK157343.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105986 | AK170681.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105986 | AK150234.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105986 | AF151985.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P105986 | NM_009304.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101632 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09674660 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09674659 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09674663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09674661 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09652801 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28167341 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28167340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28167356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09627869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167336 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09047053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09469315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167309 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09469308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09469322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09034578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167320 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09652786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09674658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167349 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167333 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09469325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09652800 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09162757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28167364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09382426 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09628240 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10382981 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382982 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382984 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382983 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417081_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448577_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10382980 | - | - |
| Mouse Genome 430 2.0 Array | 1417081_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1447638_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448577_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452944_at | - | - |
| Mouse Genome 430 2.0 Array | 1448577_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1428885_at | - | - |
| Mouse Genome 430 2.0 Array | 1452944_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417081_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1447638_at | - | - |
| Mouse Genome 430 2.0 Array | 1428885_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20973_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20973_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100018794.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018794.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018794.mm.1 | TC1100004283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018789.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018789.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018789.mm.1 | TC1100004283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018787.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018787.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018787.mm.1 | TC1100004283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018792.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018792.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018792.mm.1 | TC1100004283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018791.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018791.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018791.mm.1 | TC1100004283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018793.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018793.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018793.mm.1 | TC1100004283.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100018788.mm.1 | NM_009304.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018788.mm.1 | NM_009304.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100018788.mm.1 | TC1100004283.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417081_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448577_x_at | - | - |
| Mouse Expression Set 430 | 1448577_PM_x_at | - | - |
| Mouse Expression Set 430 | 1417081_a_at | - | - |
| Murine Genome U74v2 Set | 93586_PM_at | - | - |
| Murine Genome U74v2 Set | 93586_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448577_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448577_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417081_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417081_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417081_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448577_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448577_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417081_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 434545 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4050332 | - | - |
| MouseWG-6 v2 Expression BeadChip | 4560091 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4560091 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6130348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4050332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004050332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006130348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004560091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2706231 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1215194 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240794 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1240794 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1215194 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2706231 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001519.1_21-S | - | - |
| mouse-6 v1.1 (Illumina) | scl020973.4_189-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0020973.1_244-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4050332 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4560091 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6130348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl020973.4_189-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0020973.1_244-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0001519.1_21-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6130348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4050332 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4560091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1244874 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2706232 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2706231 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706231 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2706232 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244874 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0020973.1_244-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl020973.4_189-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0001519.1_21-S | - | - |

Syngr2
Genomic Map