Assay Details
| Gene Symbol: Sgsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197943.2 | NP_922934.2 | 11 - 12 | 1489 | 66 | |
| GenBank mRNA | AK122273.1 | - | 11 - 12 | 1411 | 66 | |
| BC058414.1 | - | 11 - 12 | 1480 | 6822416 | 66 | |
| BC060163.1 | - | 11 - 12 | 1480 | 6822416 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm2
Genomic Map