Assay Details
| Gene Symbol: Sgsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197943.2 | NP_922934.2 | 14 - 15 | 1857 | 82 | |
| GenBank mRNA | AK078057.1 | - | 3 - 4 | 1047 | 82 | |
| AK122273.1 | - | 14 - 15 | 1779 | 82 | ||
| BC058414.1 | - | 14 - 15 | 1848 | 6822416 | 82 | |
| BC060163.1 | - | 14 - 15 | 1848 | 6822416 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm2
Genomic Map