Assay Details
| Gene Symbol: Sgsm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197943.2 | NP_922934.2 | 15 - 16 | 1979 | 61 | |
| GenBank mRNA | AK078057.1 | - | 4 - 5 | 1169 | 61 | |
| AK122273.1 | - | 15 - 16 | 1901 | 61 | ||
| BC058414.1 | - | 15 - 16 | 1970 | 6822416 | 61 | |
| BC060163.1 | - | 15 - 16 | 1970 | 6822416 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgsm2
Genomic Map