| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100029516.mm.1 |
NM_197943.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029516.mm.1 |
NM_197943.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029516.mm.1 |
TC1100003204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029518.mm.1 |
NM_197943.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029518.mm.1 |
NM_197943.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029518.mm.1 |
TC1100003204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029517.mm.1 |
NM_197943.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029517.mm.1 |
NM_197943.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029517.mm.1 |
TC1100003204.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100029519.mm.1 |
NM_197943.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029519.mm.1 |
NM_197943.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100029519.mm.1 |
TC1100003204.mm.1 |
MTA TC |