Assay Details
| Gene Symbol: Pigu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004721.1 | NP_001004721.1 | 5 - 6 | 451 | 70 | |
| GenBank mRNA | AK143517.1 | - | 5 - 6 | 451 | 70 | |
| AK162913.1 | - | 426 | 70 | |||
| AK171988.1 | - | 5 - 6 | 430 | 70 | ||
| AK211799.1 | - | 3 - 4 | 231 | 70 | ||
| BC120680.1 | - | 5 - 6 | 440 | 40129603 | 70 | |
| BC120682.1 | - | 5 - 6 | 440 | 40129605 | 70 | |
| CK334159.1 | - | 5 - 6 | 404 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pigu
Genomic Map