Assay Details
| Gene Symbol: Trpc4ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163452.1 | NP_001156924.1 | 17 - 18 | 2127 | 88 | |
| NM_019828.2 | NP_062802.2 | 17 - 18 | 2103 | 88 | ||
| GenBank mRNA | AF093589.1 | - | 13 - 14 | 1599 | 88 | |
| AF130458.1 | - | 17 - 18 | 2051 | 88 | ||
| AF509881.1 | - | 17 - 18 | 2049 | 88 | ||
| AK156047.1 | - | 17 - 18 | 2076 | 88 | ||
| AK170085.1 | - | 17 - 18 | 2065 | 88 | ||
| AK171404.1 | - | 17 - 18 | 2076 | 88 | ||
| AK220239.1 | - | 17 - 18 | 2103 | 88 | ||
| BC003931.1 | - | 10 - 11 | 1063 | 88 | ||
| BC033274.1 | - | 17 - 18 | 2055 | 3673725 | 88 | |
| BC057330.1 | - | 17 - 18 | 2085 | 6816155 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trpc4ap
Genomic Map