Assay Details
| Gene Symbol: Trp53inp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178111.3 | NP_835212.1 | 1 - 2 | 164 | 78 | |
| GenBank mRNA | AJ297793.1 | - | 1 - 2 | 180 | 78 | |
| AK146589.1 | - | 1 - 2 | 169 | 78 | ||
| BC043086.1 | - | 1 - 2 | 164 | 6417598 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ297793.1, AK146589.1, BC043086.1, NM_178111.3 transcripts.
Set Membership: |
|

Trp53inp2
Genomic Map