Assay Details
| Gene Symbol: Ctnnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025680.4 | NP_079956.3 | 11 - 12 | 1337 | 68 | |
| GenBank mRNA | AK010547.1 | - | 11 - 12 | 1248 | 68 | |
| AK044690.1 | - | 11 - 12 | 1277 | 68 | ||
| AK077616.1 | - | 11 - 12 | 1281 | 68 | ||
| AK145147.1 | - | 11 - 12 | 1293 | 68 | ||
| AK145598.1 | - | 11 - 12 | 1281 | 68 | ||
| AK145719.1 | - | 11 - 12 | 1281 | 68 | ||
| AK145888.1 | - | 11 - 12 | 1298 | 68 | ||
| AK146274.1 | - | 11 - 12 | 1297 | 68 | ||
| AK206067.1 | - | 1 - 2 | 161 | 68 | ||
| BC038332.1 | - | 5 - 6 | 68 | |||
| BC050787.1 | - | 11 - 12 | 1307 | 6705648 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctnnbl1
Genomic Map