Assay Details
| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019642.4 | NP_062616.2 | 8 - 9 | 1207 | 91 | |
| GenBank mRNA | AK004968.1 | - | 8 - 9 | 1208 | 91 | |
| AK018761.2 | - | 8 - 9 | 1247 | 91 | ||
| AK087923.1 | - | 2 - 3 | 244 | 91 | ||
| AK153952.1 | - | 8 - 9 | 1068 | 91 | ||
| AK161624.1 | - | 8 - 9 | 1018 | 91 | ||
| AK166778.1 | - | 8 - 9 | 993 | 91 | ||
| AK167864.1 | - | 8 - 9 | 987 | 91 | ||
| AK167866.1 | - | 8 - 9 | 987 | 91 | ||
| AK168221.1 | - | 8 - 9 | 987 | 91 | ||
| BC010509.1 | - | 7 - 8 | 980 | 91 | ||
| BC046806.1 | - | 8 - 9 | 1235 | 6829209 | 91 | |
| D31717.1 | - | 7 - 8 | 993 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rpn2
Genomic Map