Assay Details
| Gene Symbol: Sla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029983.5 | NP_084259.1 | 2 - 3 | 314 | 102 | |
| GenBank mRNA | AF287467.1 | - | 2 - 3 | 365 | 102 | |
| AF434990.1 | - | 2 - 3 | 357 | 102 | ||
| AK030877.1 | - | 2 - 3 | 315 | 102 | ||
| AK088672.1 | - | 2 - 3 | 441 | 102 | ||
| AK138709.1 | - | 2 - 3 | 315 | 102 | ||
More Information
Set Membership: |
|

Sla2
Genomic Map