Assay Details
| Gene Symbol: Sla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029983.5 | NP_084259.1 | 5 - 6 | 608 | 117 | |
| GenBank mRNA | AF287467.1 | - | 5 - 6 | 659 | 117 | |
| AF434990.1 | - | 5 - 6 | 651 | 117 | ||
| AK020837.1 | - | 3 - 4 | 265 | 117 | ||
| AK088672.1 | - | 5 - 6 | 735 | 117 | ||
| AK138709.1 | - | 5 - 6 | 609 | 117 | ||
| BC052655.1 | - | 5 - 6 | 707 | 30040401 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | AK138709.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | BC052655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | AK030877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | AK088672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | NM_029983.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | AK138709.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | BC052655.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | AK030877.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | AK088672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P56682 | NM_029983.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252103 | AK088672.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252103 | AK088672.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503253 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 563277 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668028 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513704 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579551 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95509 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131804 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252103 | AK088672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252103 | AK088672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | AK138709.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | BC052655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | AK030877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | AK088672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | NM_029983.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | AK138709.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | BC052655.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | AK030877.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | AK088672.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P56682 | NM_029983.5 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131804 | - | - |
| Whole Mouse Genome G4122A | A_52_P56682 | AK138709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | BC052655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | AK030877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | AK088672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | NM_029983.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | AK138709.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | BC052655.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | AK030877.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | AK088672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P56682 | NM_029983.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252103 | AK088672.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P252103 | AK088672.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02062313 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02062309 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02018044 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02062318 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02062331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21773868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01554696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01795349 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773869 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01826751 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01565783 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02142334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773891 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773963 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773883 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02062309 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02048502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02142331 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01554691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01565771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01232785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773960 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773953 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01463081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773881 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02062332 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773961 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01291698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01391556 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02062319 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773923 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01565774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773968 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773911 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01565772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21773888 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489061 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489060 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489056 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489057 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489064 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489062 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489059 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489058 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489063 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437504_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451492_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477929 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489055 | - | - |
| Mouse Genome 430 2.0 Array | 1437504_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451492_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451492_at | - | - |
| Mouse Genome 430 2.0 Array | 1437504_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77799_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77799_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200042742.mm.1 | NM_029983.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042742.mm.1 | NM_029983.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042742.mm.1 | TC0200005003.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042741.mm.1 | NM_029983.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042741.mm.1 | NM_029983.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042741.mm.1 | TC0200005003.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200042740.mm.1 | NM_029983.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042740.mm.1 | NM_029983.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200042740.mm.1 | TC0200005003.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451492_PM_at | - | - |
| Mouse Expression Set 430 | 1451492_at | - | - |
| Mouse Expression Set 430 | 1437504_PM_at | - | - |
| Mouse Expression Set 430 | 1437504_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451492_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451492_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451492_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451492_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437504_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437504_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 633266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102470632 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1252407 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236595 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252407 | - | - |

Sla2
Genomic Map