Assay Details
| Gene Symbol: Ssh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291190.1 | NP_001278119.1 | 1 - 2 | 273 | 77 | |
| GenBank mRNA | AK155232.1 | - | 1 - 2 | 264 | 77 | |
| AK158449.1 | - | 1 - 2 | 274 | 77 | ||
| AK170142.1 | - | 1 - 2 | 267 | 77 | ||
More Information
Set Membership: |
|

Ssh2
Genomic Map