Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026428.2 | NP_080704.2 | 4 - 5 | 405 | 66 | |
| GenBank mRNA | AK002803.1 | - | 4 - 5 | 375 | 66 | |
| AK004023.1 | - | 4 - 5 | 404 | 66 | ||
| AK007627.1 | - | 4 - 5 | 417 | 66 | ||
| AK153521.1 | - | 4 - 5 | 386 | 66 | ||
| AK161732.1 | - | 361 | 66 | |||
| D89656.1 | - | 4 - 5 | 353 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcxr
Genomic Map