Assay Details
| Gene Symbol: Dcxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026428.2 | NP_080704.2 | 5 - 6 | 503 | 70 | |
| GenBank mRNA | AK004023.1 | - | 5 - 6 | 502 | 70 | |
| AK007627.1 | - | 5 - 6 | 515 | 70 | ||
| AK153521.1 | - | 5 - 6 | 484 | 70 | ||
| AK161732.1 | - | 459 | 70 | |||
| D89656.1 | - | 5 - 6 | 451 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcxr
Genomic Map