Assay Details
| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289738.1 | NP_001276667.1 | 1 - 2 | 136 | 139 | |
| NM_177656.4 | NP_808324.1 | 1 - 2 | 136 | 139 | ||
| NR_110367.1 | 1 - 2 | 136 | 139 | |||
| NR_110368.1 | 1 - 2 | 136 | 139 | |||
| GenBank mRNA | AK078482.1 | - | 1 - 2 | 136 | 139 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK078482.1, NM_001289738.1, NM_177656.4 transcripts.
Set Membership: |
|

6820408C15Rik
Genomic Map