Assay Details
| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177656.4 | NP_808324.1 | 4 - 5 | 587 | 86 | |
| NR_110367.1 | 4 - 5 | 587 | 86 | |||
| NR_110368.1 | 4 - 5 | 587 | 86 | |||
| GenBank mRNA | AK078482.1 | - | 4 - 5 | 587 | 86 | |
| BC144943.1 | - | 3 - 4 | 464 | 86 | ||
| BC144945.1 | - | 3 - 4 | 460 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

6820408C15Rik
Genomic Map