Assay Details
| Gene Symbol: 6820408C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289738.1 | NP_001276667.1 | 7 - 8 | 969 | 66 | |
| NM_177656.4 | NP_808324.1 | 7 - 8 | 966 | 66 | ||
| NR_110367.1 | 7 - 8 | 966 | 66 | |||
| GenBank mRNA | AK078482.1 | - | 7 - 8 | 966 | 66 | |
| BC137826.1 | - | 6 - 7 | 846 | 8860845 | 66 | |
| BC144943.1 | - | 6 - 7 | 843 | 66 | ||
| BC144944.1 | - | 6 - 7 | 846 | 9053492 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

6820408C15Rik
Genomic Map