Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081045.1 | NP_001074514.1 | 2 - 3 | 1939 | 61 | |
| GenBank mRNA | AK037800.1 | - | 3 - 4 | 1505 | 61 | |
| AK122484.1 | - | 2 - 3 | 1945 | 61 | ||
| BC043121.1 | - | 2 - 3 | 1488 | 6409084 | 61 | |
| BC053389.1 | - | 2 - 3 | 1489 | 30061629 | 61 | |
| BC054752.1 | - | 3 - 4 | 1762 | 6834541 | 61 | |
| BC079594.1 | - | 2 - 3 | 1487 | 6827379 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kansl1
Genomic Map