Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK037800.1 | - | 2 - 3 | 135 | 80 | |
| BC043121.1 | - | 1 - 2 | 118 | 6409084 | 80 | |
| BC053389.1 | - | 1 - 2 | 119 | 30061629 | 80 | |
| BC054752.1 | - | 2 - 3 | 392 | 6834541 | 80 | |
| BC079594.1 | - | 1 - 2 | 117 | 6827379 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037800.1, BC043121.1, BC053389.1, BC054752.1, BC079594.1 transcripts.
Set Membership: |
|

Kansl1
Genomic Map