Assay Details
| Gene Symbol: Prkar1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021880.2 | NP_068680.1 | 5 - 6 | 638 | 110 | |
| GenBank mRNA | AK005039.1 | - | 5 - 6 | 640 | 110 | |
| AK019147.1 | - | 3 - 4 | 273 | 110 | ||
| AK027916.1 | - | 5 - 6 | 605 | 110 | ||
| AK049832.1 | - | 5 - 6 | 631 | 110 | ||
| AK051068.1 | - | 5 - 6 | 639 | 110 | ||
| AK053918.1 | - | 5 - 6 | 639 | 110 | ||
| AK083383.1 | - | 5 - 6 | 619 | 110 | ||
| AK145797.1 | - | 5 - 6 | 639 | 110 | ||
| AK145860.1 | - | 5 - 6 | 606 | 110 | ||
| AK146380.1 | - | 5 - 6 | 625 | 110 | ||
| AK147188.1 | - | 5 - 6 | 630 | 110 | ||
| AK150427.1 | - | 5 - 6 | 631 | 110 | ||
| AK150927.1 | - | 3 - 4 | 271 | 110 | ||
| AK151040.1 | - | 3 - 4 | 271 | 110 | ||
| AK151124.1 | - | 5 - 6 | 597 | 110 | ||
| AK151556.1 | - | 3 - 4 | 271 | 110 | ||
| AK152044.1 | - | 3 - 4 | 271 | 110 | ||
| AK153227.1 | - | 5 - 6 | 597 | 110 | ||
| AK158562.1 | - | 5 - 6 | 976 | 110 | ||
| BC003461.1 | - | 5 - 6 | 609 | 2648357 | 110 | |
| BC005697.1 | - | 5 - 6 | 580 | 3597765 | 110 | |
| HQ412666.1 | - | 6 - 7 | 821 | 110 | ||
| HQ412667.1 | - | 5 - 6 | 637 | 110 | ||
| HQ412668.1 | - | 5 - 6 | 697 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkar1a
Genomic Map