Assay Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145823.2 | NP_665822.1 | 5 - 6 | 1141 | 89 | |
| GenBank mRNA | AB077281.1 | - | 5 - 6 | 1122 | 89 | |
| AB077282.1 | - | 5 - 6 | 419 | 89 | ||
| AK003844.1 | - | 4 - 5 | 389 | 89 | ||
| AK021206.1 | - | 5 - 6 | 354 | 89 | ||
| AK153625.1 | - | 5 - 6 | 1142 | 89 | ||
| AK171340.1 | - | 5 - 6 | 1168 | 89 | ||
| AK171918.1 | - | 5 - 6 | 1138 | 89 | ||
| BC082333.1 | - | 5 - 6 | 973 | 89 | ||
| BC108351.1 | - | 5 - 6 | 1001 | 6312193 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnc1
Genomic Map