Assay Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145823.2 | NP_665822.1 | 6 - 7 | 1242 | 78 | |
| GenBank mRNA | AB077281.1 | - | 6 - 7 | 1223 | 78 | |
| AB077282.1 | - | 6 - 7 | 520 | 78 | ||
| AK003844.1 | - | 5 - 6 | 490 | 78 | ||
| AK021206.1 | - | 6 - 7 | 455 | 78 | ||
| AK153625.1 | - | 6 - 7 | 1243 | 78 | ||
| AK171340.1 | - | 6 - 7 | 1269 | 78 | ||
| AK171918.1 | - | 6 - 7 | 1239 | 78 | ||
| BC082333.1 | - | 6 - 7 | 1074 | 78 | ||
| BC108351.1 | - | 6 - 7 | 1102 | 6312193 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pitpnc1
Genomic Map