Assay Details
| Gene Symbol: Rbm39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291114.1 | NP_001278043.1 | 9 - 10 | 1193 | 127 | |
| NM_001291115.1 | NP_001278044.1 | 9 - 10 | 988 | 127 | ||
| NM_133242.3 | NP_573505.2 | 9 - 10 | 1193 | 127 | ||
| GenBank mRNA | AK135289.1 | - | 8 - 9 | 819 | 127 | |
| AK146046.1 | - | 9 - 10 | 988 | 127 | ||
| AK147076.1 | - | 9 - 10 | 1138 | 127 | ||
| AK154989.1 | - | 9 - 10 | 1124 | 127 | ||
| BC004000.1 | - | 5 - 6 | 525 | 127 | ||
| BC023205.1 | - | 9 - 10 | 1095 | 127 | ||
| BC030493.1 | - | 7 - 8 | 808 | 5400675 | 127 | |
| BC082607.1 | - | 7 - 8 | 783 | 30667785 | 127 | |
| BC086645.1 | - | 9 - 10 | 1116 | 6817169 | 127 | |
| BC092268.1 | - | 7 - 8 | 942 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm39
Genomic Map