Assay Details
| Gene Symbol: Vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019806.5 | NP_062780.2 | 3 - 4 | 438 | 94 | |
| GenBank mRNA | AF115504.1 | - | 3 - 4 | 316 | 94 | |
| AK050632.1 | - | 3 - 4 | 387 | 94 | ||
| AK081908.1 | - | 3 - 4 | 367 | 94 | ||
| AK160925.1 | - | 3 - 4 | 281 | 94 | ||
| AK172080.1 | - | 3 - 4 | 280 | 94 | ||
| BC048231.1 | - | 3 - 4 | 438 | 6313869 | 94 | |
| BC057123.1 | - | 3 - 4 | 427 | 6411688 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vapb
Genomic Map