Assay Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024436.3 | NP_077756.2 | 2 - 3 | 372 | 74 | |
| GenBank mRNA | AB232616.1 | - | 2 - 3 | 111 | 74 | |
| AJ311124.1 | - | 2 - 3 | 168 | 74 | ||
| AK014407.1 | - | 358 | 74 | |||
| AK031667.1 | - | 2 - 3 | 352 | 74 | ||
| AK146594.1 | - | 2 - 3 | 372 | 74 | ||
| AK149200.1 | - | 364 | 74 | |||
| AK150617.1 | - | 2 - 3 | 378 | 74 | ||
| BC006596.1 | - | 2 - 3 | 359 | 3965264 | 74 | |
| M79304.1 | - | 1 - 2 | 63 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab22a
Genomic Map