Assay Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024436.3 | NP_077756.2 | 4 - 5 | 530 | 71 | |
| GenBank mRNA | AB232616.1 | - | 4 - 5 | 269 | 71 | |
| AJ311124.1 | - | 4 - 5 | 326 | 71 | ||
| AK031667.1 | - | 4 - 5 | 510 | 71 | ||
| AK146594.1 | - | 4 - 5 | 530 | 71 | ||
| AK147619.1 | - | 534 | 71 | |||
| AK149200.1 | - | 522 | 71 | |||
| AK150617.1 | - | 4 - 5 | 536 | 71 | ||
| BC006596.1 | - | 4 - 5 | 517 | 3965264 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab22a
Genomic Map