Assay Details
| Gene Symbol: Cct2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007636.2 | NP_031662.2 | 6 - 7 | 555 | 74 | |
| GenBank mRNA | AB041570.1 | - | 6 - 7 | 527 | 74 | |
| AK012295.1 | - | 6 - 7 | 511 | 74 | ||
| AK075581.1 | - | 6 - 7 | 528 | 74 | ||
| AK088145.1 | - | 6 - 7 | 517 | 74 | ||
| AK145406.1 | - | 6 - 7 | 556 | 74 | ||
| AK166891.1 | - | 6 - 7 | 528 | 74 | ||
| BC007470.1 | - | 6 - 7 | 519 | 3601569 | 74 | |
| BC026918.1 | - | 6 - 7 | 521 | 4500896 | 74 | |
| Z31553.1 | - | 6 - 7 | 506 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cct2
Genomic Map