Assay Details
| Gene Symbol: Mgat4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026233.2 | NP_080509.2 | 6 - 7 | 695 | 123 | |
| GenBank mRNA | AK016272.1 | - | 6 - 7 | 704 | 123 | |
| AK017057.1 | - | 3 - 4 | 285 | 123 | ||
| HM067443.1 | - | 6 - 7 | 819 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgat4d
Genomic Map